Assay Details
| Gene Symbol: Pold4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027196.3 | NP_081472.1 | 3 - 4 | 490 | 90 | |
| GenBank mRNA | AF515709.1 | - | 3 - 4 | 390 | 90 | |
| AK010477.1 | - | 3 - 4 | 453 | 90 | ||
| AK170469.1 | - | 2 - 3 | 226 | 90 | ||
| BC028520.1 | - | 3 - 4 | 411 | 1400386 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774814 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921960 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09090116 | - | - |
| Whole Mouse Genome G4122A | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | NM_027196.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | AF515709.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | AK010477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | AK170469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P455906 | BC028520.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102858 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134671 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14297098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14297059 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14297089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14297094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31922031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14279794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14278528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14207171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14341857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14298777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14297055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14297083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14222589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14297059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14207166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14470748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14501555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14470747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14297066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31922000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14470741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14470754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14297058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14501557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460394 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427885_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460392 | - | - |
| Mouse Genome 430 2.0 Array | 1427885_at | - | - |
| Mouse Genome 430 2.0 Array | 1427885_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69745_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69745_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000509.mm.1 | NM_027196.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000509.mm.1 | TC1900000036.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427885_at | - | - |
| Mouse Expression Set 430 | 1427885_PM_at | - | - |
| Murine Genome U74v2 Set | 94843_at | - | - |
| Murine Genome U74v2 Set | 94843_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427885_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427885_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427885_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427885_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 924237 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2320082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777247 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777247 | - | - |
| mouse-6 v1.1 (Illumina) | scl069745.1_205-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320082 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl069745.1_205-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2910596 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2910596 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2777247 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl069745.1_205-S | - | - |

Pold4
Genomic Map