Assay Details
| Gene Symbol: Trmt10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027266.4 | NP_081542.2 | 5 - 6 | 636 | 92 | |
| GenBank mRNA | AK011746.1 | - | 5 - 6 | 649 | 92 | |
| AK030465.1 | - | 5 - 6 | 637 | 92 | ||
| BC019428.1 | - | 4 - 5 | 613 | 4205772 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt10b
Genomic Map