Assay Details
| Gene Symbol: Ppil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285826.1 | NP_001272755.1 | 2 - 3 | 733 | 103 | |
| NM_001285827.1 | NP_001272756.1 | 2 - 3 | 398 | 103 | ||
| NM_027351.3 | NP_081627.1 | 2 - 3 | 212 | 103 | ||
| NM_027374.3 | NP_081650.2 | 2 - 3 | 398 | 103 | ||
| GenBank mRNA | AK010207.1 | - | 2 - 3 | 216 | 103 | |
| AK011000.1 | - | 2 - 3 | 167 | 103 | ||
| BC060993.1 | - | 2 - 3 | 180 | 6591278 | 103 | |
| BC061645.1 | - | 2 - 3 | 676 | 5032617 | 103 | |
| BY288296.1 | - | 2 - 3 | 339 | 103 | ||
| CJ166559.1 | - | 2 - 3 | 342 | 103 | ||
| CJ170065.1 | - | 2 - 3 | 342 | 103 | ||
| DV043111.1 | - | 2 - 3 | 434 | 103 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK010207.1,AK011000.1,BC060993.1,BC061645.1,NM_001285826.1,NM_001285827.1,NM_027351.3,NM_027374.3 transcripts.
Set Membership: |
|

Ppil3
Genomic Map