Assay Details
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027541.4 | NP_081817.2 | 5 - 6 | 991 | 90 | |
GenBank mRNA | AK014398.1 | - | 5 - 6 | 599 | 90 | |
AK088194.1 | - | 5 - 6 | 992 | 90 | ||
AK139852.1 | - | 5 - 6 | 624 | 90 | ||
AK167435.1 | - | 5 - 6 | 628 | 90 | ||
BC026607.1 | - | 2 - 3 | 745 | 90 | ||
BC040787.1 | - | 2 - 3 | 745 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -




Gene Ontology Categories:
MicroArray Probes:
