Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500030875.mm.1 |
TC0500003414.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030875.mm.1 |
NM_027891.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030875.mm.1 |
NM_027891.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030872.mm.1 |
TC0500003414.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030872.mm.1 |
NM_027891.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030872.mm.1 |
NM_027891.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030873.mm.1 |
TC0500003414.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030873.mm.1 |
NM_027891.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030873.mm.1 |
NM_027891.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030878.mm.1 |
TC0500003414.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030878.mm.1 |
NM_027891.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030878.mm.1 |
NM_027891.4 |
Refseq |