Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400017555.mm.1 |
NM_028020.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017555.mm.1 |
NM_028020.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017555.mm.1 |
TC0400002097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017554.mm.1 |
NM_028020.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017554.mm.1 |
NM_028020.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017554.mm.1 |
TC0400002097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017553.mm.1 |
NM_028020.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017553.mm.1 |
NM_028020.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017553.mm.1 |
TC0400002097.mm.1 |
MTA TC |