Assay Details
| Gene Symbol: Trmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028063.2 | NP_082339.1 | 8 - 9 | 920 | 79 | |
| GenBank mRNA | AK005541.1 | - | 8 - 9 | 920 | 79 | |
| AK043532.1 | - | 8 - 9 | 927 | 79 | ||
| AK044059.1 | - | 8 - 9 | 927 | 79 | ||
| AY349617.1 | - | 8 - 9 | 927 | 79 | ||
| BC037371.1 | - | 2 - 3 | 175 | 79 | ||
| BC051548.1 | - | 8 - 9 | 921 | 79 | ||
| BC061026.1 | - | 8 - 9 | 907 | 30305575 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmu
Genomic Map