Assay Details
| Gene Symbol: Slc39a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028064.2 | NP_082340.1 | 10 - 11 | 1757 | 69 | |
| GenBank mRNA | AB052762.1 | - | 4 - 5 | 376 | 69 | |
| AK005535.1 | - | 10 - 11 | 1797 | 69 | ||
| AK146977.1 | - | 10 - 11 | 1798 | 69 | ||
| AK147107.1 | - | 10 - 11 | 1776 | 69 | ||
| AK154089.1 | - | 10 - 11 | 2276 | 69 | ||
| AK155467.1 | - | 10 - 11 | 2276 | 69 | ||
| BC023498.1 | - | 10 - 11 | 1757 | 5011088 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a4
Genomic Map