Assay Details
| Gene Symbol: Gcnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028087.2 | NP_082363.2 | 2 - 3 | 464 | 79 | |
| GenBank mRNA | AK008234.1 | - | 1 - 2 | 74 | 79 | |
| AY039027.1 | - | 1 - 2 | 138 | 79 | ||
| EF202835.1 | - | 2 - 3 | 464 | 79 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008234.1,AY039027.1,EF202835.1,NM_028087.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immunoglobulin production in mucosal tissue
protein O-linked glycosylation
tissue morphogenesis
intestinal absorption
kidney morphogenesis
beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity
N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase activity
acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity
immunoglobulin production in mucosal tissue
protein O-linked glycosylation
tissue morphogenesis
intestinal absorption
kidney morphogenesis
beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity
N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase activity
acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11011 | EF202835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11011 | AK008789.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11011 | NM_028087.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118861 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132259 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11011 | EF202835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11011 | AK008789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11011 | NM_028087.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26614327 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132259 | - | - |
| Whole Mouse Genome G4122A | A_52_P53144 | BC018297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P53144 | EF202835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P53144 | AK008234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P53144 | AY039027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P53144 | NM_028087.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103843 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132259 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07671238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08074001 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07768926 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08074004 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08074000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08073999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26614270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08074001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08007658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07767076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08007645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08074000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08007648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07948331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26614319 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10594760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10594759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10594761 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430963_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440409_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424901_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10594758 | - | - |
| Mouse Genome 430 2.0 Array | 1430963_at | - | - |
| Mouse Genome 430 2.0 Array | 1430963_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424901_at | - | - |
| Mouse Genome 430 2.0 Array | 1424901_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72077_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72077_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900021532.mm.1 | NM_028087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021532.mm.1 | NM_028087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021532.mm.1 | TC0900002586.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900021528.mm.1 | NM_028087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021528.mm.1 | NM_028087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021528.mm.1 | TC0900002586.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900021529.mm.1 | NM_028087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021529.mm.1 | NM_028087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021529.mm.1 | TC0900002586.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900021531.mm.1 | NM_028087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021531.mm.1 | NM_028087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021531.mm.1 | TC0900002586.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900021535.mm.1 | NM_028087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021535.mm.1 | NM_028087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021535.mm.1 | TC0900002586.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900021530.mm.1 | NM_028087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021530.mm.1 | NM_028087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021530.mm.1 | TC0900002586.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900021534.mm.1 | NM_028087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021534.mm.1 | NM_028087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900021534.mm.1 | TC0900002586.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424901_PM_at | - | - |
| Mouse Expression Set 430 | 1424901_at | - | - |
| Mouse Expression Set 430 | 1430963_at | - | - |
| Mouse Expression Set 430 | 1430963_PM_at | - | - |
| Murine Genome U74v2 Set | 130647_at | - | - |
| Murine Genome U74v2 Set | 132809_PM_at | - | - |
| Murine Genome U74v2 Set | 132809_at | - | - |
| Murine Genome U74v2 Set | 130647_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424901_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424901_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424901_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424901_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440409_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430963_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430963_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440409_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 360361 | - | - |
| ABI Mouse Genome Survey Microarray | 830600 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6290239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250994 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250994 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2445378 | - | - |
| mouse-6 v1.1 (Illumina) | scl072077.1_53-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl072077.1_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290239 | - | - |

Gcnt3
Genomic Map