Assay Details
| Gene Symbol: Jmjd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028101.4 | NP_082377.2 | 6 - 7 | 555 | 56 | |
| GenBank mRNA | AK011308.1 | - | 6 - 7 | 552 | 56 | |
| AK148079.1 | - | 6 - 7 | 533 | 56 | ||
| AK155810.1 | - | 6 - 7 | 557 | 56 | ||
| AK172073.1 | - | 6 - 7 | 552 | 56 | ||
| BC010800.1 | - | 6 - 7 | 517 | 4235072 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P434776 | AK011308.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P434776 | AK011308.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55902 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175261 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P434776 | AK011308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P434776 | AK011308.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106556 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175261 | - | - |
| Whole Mouse Genome G4122A | A_51_P434776 | AK011308.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434776 | AK011308.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127503 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13128245 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13128243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13351714 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13128248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13351714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13128247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106547 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442907 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449053 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442909 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449052 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449054 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442905 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424743_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442904 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449051 | - | - |
| Mouse Genome 430 2.0 Array | 1424743_at | - | - |
| Mouse Genome 430 2.0 Array | 1424743_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72106_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72106_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700003311.mm.1 | NM_028101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003311.mm.1 | NM_028101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003311.mm.1 | TC1700000391.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424743_PM_at | - | - |
| Mouse Expression Set 430 | 1424743_at | - | - |
| Murine Genome U74v2 Set | 94520_at | - | - |
| Murine Genome U74v2 Set | 94520_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424743_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424743_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424743_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424743_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 485403 | - | - |
| ABI Mouse Genome Survey Microarray | 759175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6450575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6450575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006450575 | - | - |
| mouse-6 v1.1 (Illumina) | scl072106.9_198-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6450575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl072106.9_198-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6450575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2896492 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2896492 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl072106.9_198-S | - | - |

Jmjd8
Genomic Map