Assay Details
| Gene Symbol: Faim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028224.4 | NP_082500.2 | 2 - 3 | 788 | 83 | |
| GenBank mRNA | AF468028.1 | - | 1 - 2 | 217 | 83 | |
| AK129246.1 | - | 2 - 3 | 301 | 83 | ||
| AK147343.1 | - | 2 - 3 | 306 | 83 | ||
| AK158960.1 | - | 2 - 3 | 581 | 83 | ||
| BC072560.1 | - | 2 - 3 | 788 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faim2
Genomic Map