Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100006355.mm.1 |
NM_028360.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006355.mm.1 |
NM_028360.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006355.mm.1 |
NM_029704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006355.mm.1 |
NM_029704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006355.mm.1 |
TC1100000794.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006353.mm.1 |
NM_028360.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006353.mm.1 |
NM_028360.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006353.mm.1 |
NM_029704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006353.mm.1 |
NM_029704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006353.mm.1 |
TC1100000794.mm.1 |
MTA TC |