Assay Details
| Gene Symbol: 1700074P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028550.3 | NP_082826.1 | 5 - 6 | 819 | 115 | |
| GenBank mRNA | AK006951.1 | - | 4 - 5 | 693 | 115 | |
| AK030264.1 | - | 5 - 6 | 819 | 115 | ||
| BC061035.1 | - | 4 - 5 | 730 | 6774253 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100451 | AK030264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100451 | BC061035.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100451 | NM_028550.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100451 | AK006951.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899096 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178946 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133634 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105775 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100451 | AK030264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100451 | BC061035.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100451 | NM_028550.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100451 | AK006951.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391641 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05875040 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05875025 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05875068 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133634 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105775 | - | - |
| Whole Mouse Genome G4122A | A_51_P469510 | AK030264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469510 | BC061035.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469510 | NM_028550.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469510 | AK006951.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100451 | AK030264.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100451 | BC061035.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100451 | NM_028550.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100451 | AK006951.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107045 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133634 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105775 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05108153 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05108148 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05108136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05108140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05108139 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05108149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05239207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05364687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05108144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05880084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05494374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05880086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05880091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05201397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05364686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05686301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05201395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05421890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24391645 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10544310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544313 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544311 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544312 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544314 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432301_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10544307 | - | - |
| Mouse Genome 430 2.0 Array | 1432301_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432301_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73481_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73481_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600016946.mm.1 | TC0600002147.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600016946.mm.1 | NM_028550.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600016946.mm.1 | NM_028550.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600016947.mm.1 | TC0600002147.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600016947.mm.1 | NM_028550.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600016947.mm.1 | NM_028550.3 | Refseq |
| Mouse Expression Set 430 | 1432301_PM_a_at | - | - |
| Mouse Expression Set 430 | 1432301_a_at | - | - |
| Murine Genome U74v2 Set | 101839_PM_at | - | - |
| Murine Genome U74v2 Set | 101839_at | - | - |
| Murine Genome U74v2 Set | 167541_r_at | - | - |
| Murine Genome U74v2 Set | 167541_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432301_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432301_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432301_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432301_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 368387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5670161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005670161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2695988 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2695988 | - | - |
| mouse-6 v1.1 (Illumina) | scl29090.7_52-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5670161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29090.7_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5670161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2695988 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2695988 | - | - |

1700074P13Rik
Genomic Map