Assay Details
Gene Symbol: Fam122c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028671.2 | NP_082947.1 | 1 - 2 | 65 | 117 | |
GenBank mRNA | AK015288.1 | - | 1 - 2 | 65 | 117 | |
AK015916.1 | - | 1 - 2 | 75 | 117 | ||
BC147640.1 | - | 1 - 2 | 53 | 9056630 | 117 | |
BC147646.1 | - | 1 - 2 | 53 | 9056636 | 117 | |
BC172114.1 | - | 1 - 2 | 53 | 9054400 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK015288.1, AK015916.1, BC147640.1, BC147646.1, BC172114.1, NM_028671.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
