Assay Details
| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028716.4 | NP_082992.1 | 4 - 5 | 405 | 119 | |
| GenBank mRNA | AK014030.1 | - | 3 - 4 | 825 | 119 | |
| AK014380.1 | - | 4 - 5 | 406 | 119 | ||
| BC027776.1 | - | 4 - 5 | 400 | 5352251 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf19
Genomic Map