Assay Details
| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028716.4 | NP_082992.1 | 13 - 14 | 1346 | 99 | |
| GenBank mRNA | AK014380.1 | - | 13 - 14 | 1347 | 99 | |
| BC027776.1 | - | 13 - 14 | 1341 | 5352251 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf19
Genomic Map