Assay Details
Gene Symbol: Pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014598.1 | - | 4 - 5 | 1194 | 69 | |
AK040411.1 | - | 1184 | 69 | |||
AK087637.1 | - | 4 - 5 | 1416 | 69 | ||
AK145961.1 | - | 3 - 4 | 899 | 69 | ||
AK147925.1 | - | 4 - 5 | 1186 | 69 | ||
BC019144.1 | - | 4 - 5 | 1177 | 5050939 | 69 | |
BC058694.1 | - | 4 - 5 | 1167 | 6811976 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981210 | AK087637.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981210 | AK042663.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981210 | AK014598.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981210 | NM_028730.5 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969660 | BC058694.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969660 | BC019144.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969660 | AK147925.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969660 | AK149594.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969660 | AK145961.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69908 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727304 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776548 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683309 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46294 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200608 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823289 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172660 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87867 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981210 | AK087637.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981210 | AK042663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981210 | AK014598.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981210 | NM_028730.5 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969660 | BC058694.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969660 | BC019144.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969660 | AK147925.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969660 | AK149594.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969660 | AK145961.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P354306 | BC058694.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P354306 | BC019144.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P354306 | AK147925.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P354306 | AK149594.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P354306 | AK145961.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121974 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128662 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10541294 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541296 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541293 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541292 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541295 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541297 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541298 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541299 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541291 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451393_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10541290 | - | - |
Mouse Genome 430 2.0 Array | 1451393_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451393_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74043_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74043_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600010858.mm.1 | TC0600001366.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010858.mm.1 | NM_028730.6 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010861.mm.1 | TC0600001366.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010861.mm.1 | NM_028730.6 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010860.mm.1 | TC0600001366.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010860.mm.1 | NM_028730.6 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010862.mm.1 | TC0600001366.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010862.mm.1 | NM_028730.6 | Refseq |
Mouse Expression Set 430 | 1451393_at | - | - |
Mouse Expression Set 430 | 1451393_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451393_at | - | - |
Mouse Genome 430A 2.0 Array | 1451393_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451393_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451393_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 671220 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5890537 | - | - |
MouseWG-6 v2 Expression BeadChip | 102100397 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106450673 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102100397 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5890537 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005890537 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652077 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1221450 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2438370 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2652077 | - | - |
mouse-6 v1.1 (Illumina) | scl0074043.2_32-S | - | - |
mouse-6 v1.1 (Illumina) | scl29556.6.1_40-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102100397 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5890537 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106450673 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0074043.2_32-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890537 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106450673 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102100397 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2652077 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2652077 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2438370 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221450 | - | - |
