Assay Details
Gene Symbol: 4933404G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045920.1 | 2 - 3 | 186 | 96 | ||
GenBank mRNA | AK016605.1 | - | 2 - 3 | 164 | 96 | |
AK016649.1 | - | 2 - 3 | 162 | 96 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK016649.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AK016605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AK016649.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AV270515.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AV278869.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AK016605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AK016649.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AV270515.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P367374 | AV278869.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963495 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293185 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48584 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AK016605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AK016649.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AV270515.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AV278869.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AK016605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AK016649.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AV270515.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P367374 | AV278869.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963495 | - | - |
Whole Mouse Genome G4122A | A_51_P367374 | AK016605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P367374 | AK016649.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P367374 | AV270515.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P367374 | AV278869.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P367374 | AK016605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P367374 | AK016649.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P367374 | AV270515.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P367374 | AV278869.1 | mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570259 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13000430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12628746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12772672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570258 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12628748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570310 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570283 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570255 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570256 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570329 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570260 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30570299 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10438048 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10438043 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10438047 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431509_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431509_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431509_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74066_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74066_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1600010244.mm.1 | NR_045920.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010244.mm.1 | NR_045920.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010244.mm.1 | TC1600001265.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431509_PM_at | - | - |
Mouse Expression Set 430 | 1431509_at | - | - |
Murine Genome U74v2 Set | 167450_PM_at | - | - |
Murine Genome U74v2 Set | 167450_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431509_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431509_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 416342 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5220725 | - | - |
