Assay Details
| Gene Symbol: 4933404G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045920.1 | 3 - 4 | 639 | 97 | ||
| GenBank mRNA | AK016649.1 | - | 3 - 4 | 615 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

4933404G15Rik
Genomic Map