Assay Details
| Gene Symbol: Acoxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028765.3 | NP_083041.1 | 8 - 9 | 1035 | 78 | |
| GenBank mRNA | AK004775.1 | - | 8 - 9 | 1037 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acoxl
Genomic Map