Assay Details
Gene Symbol: Dmgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028772.3 | NP_083048.1 | 2 - 3 | 268 | 104 | |
GenBank mRNA | AK004755.1 | - | 2 - 3 | 269 | 104 | |
BC089599.1 | - | 2 - 3 | 271 | 30118948 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
