Assay Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028772.3 | NP_083048.1 | 13 - 14 | 2180 | 84 | |
| GenBank mRNA | AK004755.1 | - | 13 - 14 | 2181 | 84 | |
| BC024126.1 | - | 9 - 10 | 1463 | 84 | ||
| BC089599.1 | - | 13 - 14 | 2183 | 30118948 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmgdh
Genomic Map