Assay Details
| Gene Symbol: Ampd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289719.1 | NP_001276648.1 | 12 - 13 | 1684 | 67 | |
| NM_001289720.1 | NP_001276649.1 | 11 - 12 | 1501 | 67 | ||
| NM_028779.5 | NP_083055.1 | 11 - 12 | 1770 | 67 | ||
| GenBank mRNA | AK004759.2 | - | 12 - 13 | 1669 | 67 | |
| AK034844.1 | - | 11 - 12 | 1475 | 67 | ||
| AK169980.1 | - | 12 - 13 | 1670 | 67 | ||
| AK187620.1 | - | 2 - 3 | 157 | 67 | ||
| BC049119.1 | - | 11 - 12 | 1731 | 6812571 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ampd2
Genomic Map