Assay Details
| Gene Symbol: Ampd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289719.1 | NP_001276648.1 | 14 - 15 | 1975 | 60 | |
| NM_001289720.1 | NP_001276649.1 | 13 - 14 | 1792 | 60 | ||
| NM_028779.5 | NP_083055.1 | 13 - 14 | 2061 | 60 | ||
| GenBank mRNA | AK004759.2 | - | 14 - 15 | 1960 | 60 | |
| AK034844.1 | - | 13 - 14 | 1766 | 60 | ||
| AK169980.1 | - | 14 - 15 | 1961 | 60 | ||
| BC016662.1 | - | 2 - 3 | 285 | 60 | ||
| BC049119.1 | - | 13 - 14 | 2022 | 6812571 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ampd2
Genomic Map