Assay Details
| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028841.3 | NP_083117.2 | 5 - 6 | 774 | 72 | |
| GenBank mRNA | AI019499.1 | - | 2 - 3 | 245 | 72 | |
| AK008761.1 | - | 5 - 6 | 719 | 72 | ||
| AK166272.1 | - | 5 - 6 | 777 | 72 | ||
| AK171691.1 | - | 5 - 6 | 723 | 72 | ||
| BC010346.1 | - | 5 - 6 | 719 | 2811935 | 72 | |
| CD740983.1 | - | 4 - 5 | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tspan17
Genomic Map