Assay Details
| Gene Symbol: Zc3hav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028421.1 | NP_082697.1 | 5 - 6 | 2220 | 77 | |
| NM_028864.2 | NP_083140.1 | 5 - 6 | 2220 | 77 | ||
| GenBank mRNA | AK004770.1 | - | 5 - 6 | 2221 | 77 | |
| AK020263.2 | - | 5 - 6 | 1899 | 77 | ||
| AK080334.1 | - | 5 - 6 | 2205 | 77 | ||
| AK143568.1 | - | 5 - 6 | 1890 | 77 | ||
| AK169402.1 | - | 5 - 6 | 1865 | 77 | ||
| AK172379.1 | - | 5 - 6 | 2221 | 77 | ||
| BC029090.1 | - | 5 - 6 | 1782 | 2651558 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zc3hav1
Genomic Map