Assay Details
| Gene Symbol: Sgms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028943.5 | NP_083219.2 | 1 - 2 | 332 | 74 | |
| GenBank mRNA | AK016659.1 | - | 1 - 2 | 346 | 74 | |
| BC117782.2 | - | 1 - 2 | 336 | 40092081 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK016659.1,BC117782.2,NM_028943.5 transcripts.
Set Membership: |
|

Sgms2
Genomic Map