Assay Details
| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028946.3 | NP_083222.2 | 4 - 5 | 889 | 80 | |
| GenBank mRNA | AK016917.1 | - | 4 - 5 | 869 | 80 | |
| AK029525.1 | - | 4 - 5 | 890 | 80 | ||
| BC025002.1 | - | 3 - 4 | 811 | 80 | ||
| EU846100.1 | - | 3 - 4 | 420 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9b1
Genomic Map