Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300007684.mm.1 |
NM_029001.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007684.mm.1 |
NM_029001.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007684.mm.1 |
TC1300001204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300007682.mm.1 |
NM_029001.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007682.mm.1 |
NM_029001.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007682.mm.1 |
TC1300001204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300007683.mm.1 |
NM_029001.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007683.mm.1 |
NM_029001.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007683.mm.1 |
TC1300001204.mm.1 |
MTA TC |