Assay Details
| Gene Symbol: Cbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028763.3 | NP_083039.2 | 5 | 3687 | 61 | |
| GenBank mRNA | AK017510.1 | - | 1 | 48 | 61 | |
| AK162544.1 | - | 1 | 687 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK017510.1,AK162544.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368806 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368846 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368841 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368807 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368805 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368845 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368843 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111741 | BC048240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111741 | BC011199.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111741 | AK146048.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111741 | AK146103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111741 | AK147702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120238 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127960 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104635 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111741 | BC048240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111741 | BC011199.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111741 | AK146048.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111741 | AK146103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111741 | AK147702.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30368868 | - | - |
| Whole Mouse Genome G4122A | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221428 | BC019942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221428 | AK162544.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221428 | AK017510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221428 | NM_028763.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392121 | BC048240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392121 | BC011199.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392121 | AK146048.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392121 | AK146103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P392121 | AK147702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | BC048240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | AK218930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | BC011199.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | AK162544.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P391764 | AK202886.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100286 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118159 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125125 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11843718 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11912216 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12095882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11912217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12095878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11843709 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368846 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368844 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368806 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368805 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368841 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368845 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368807 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30368843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430643 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429290_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424407_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430626 | - | - |
| Mouse Genome 430 2.0 Array | 1424407_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429290_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424407_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429290_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 494448_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 494448_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014852.mm.1 | NM_028763.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014852.mm.1 | NM_028763.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014852.mm.1 | TC1500001887.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424407_s_at | - | - |
| Mouse Expression Set 430 | 1421181_PM_at | - | - |
| Mouse Expression Set 430 | 1450147_at | - | - |
| Mouse Expression Set 430 | 1450147_PM_at | - | - |
| Mouse Expression Set 430 | 1421181_at | - | - |
| Mouse Expression Set 430 | 1424407_PM_s_at | - | - |
| Mouse Expression Set 430 | 1434270_at | - | - |
| Mouse Expression Set 430 | 1434270_PM_at | - | - |
| Mouse Expression Set 430 | 1429290_PM_at | - | - |
| Mouse Expression Set 430 | 1429290_at | - | - |
| Murine Genome U74v2 Set | 162138_PM_s_at | - | - |
| Murine Genome U74v2 Set | 162138_s_at | - | - |
| Murine Genome U74v2 Set | 160942_at | - | - |
| Murine Genome U74v2 Set | 160942_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424407_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424407_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450147_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450147_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421181_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421181_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 459094 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102970427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102970427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060685 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107040332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060685 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2598604 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2598604 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258688 | - | - |
| mouse-6 v1.1 (Illumina) | scl46953.1.327_25-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0073340.1_282-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107040332 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102970427 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060685 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0073340.1_282-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107040332 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102970427 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060685 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2598604 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2598604 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258688 | - | - |

Cbx6
Genomic Map