Assay Details
| Gene Symbol: Sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029157.3 | NP_083433.1 | 11 - 12 | 972 | 105 | |
| GenBank mRNA | AK030743.1 | - | 11 - 12 | 1007 | 105 | |
| AK150733.1 | - | 11 - 12 | 970 | 105 | ||
| AK151141.1 | - | 11 - 12 | 970 | 105 | ||
| AK151382.1 | - | 11 - 12 | 970 | 105 | ||
| AK161646.1 | - | 11 - 12 | 993 | 105 | ||
| AK168966.1 | - | 11 - 12 | 996 | 105 | ||
| AK169324.1 | - | 11 - 12 | 1016 | 105 | ||
| BC009141.1 | - | 11 - 12 | 972 | 3598647 | 105 | |
| BC092058.1 | - | 11 - 12 | 1004 | 4017388 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269840 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | AK216219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | BC009141.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | BC092058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | AK161646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | AK168966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | AK216219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | BC009141.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | BC092058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | AK161646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297165 | AK168966.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481820 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | AK044619.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | BC036966.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | BC040465.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | BC052329.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | BC060083.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | DQ176851.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | NM_001113360.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325825 | NM_175556.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48468 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173957 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | AK216219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | BC009141.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | BC092058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | AK161646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | AK168966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | AK216219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | BC009141.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | BC092058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | AK161646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297165 | AK168966.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173957 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05387626 | - | - |
| Whole Mouse Genome G4122A | A_51_P297165 | AK216219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | BC009141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | BC092058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | AK161646.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | AK168966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | AK216219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | BC009141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | BC092058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | AK161646.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297165 | AK168966.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132208 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100807 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108742 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173957 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318526 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318528 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318522 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23269840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03437977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03340942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03335324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03340940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03437979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03437973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03340939 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507929 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423811_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432488_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493921 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507914 | - | - |
| Mouse Genome 430 2.0 Array | 1423811_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432488_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432488_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423811_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75062_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75062_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400010687.mm.1 | NM_029157.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010687.mm.1 | NM_029157.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010687.mm.1 | TC0400001421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010688.mm.1 | NM_029157.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010688.mm.1 | NM_029157.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010688.mm.1 | TC0400001421.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432488_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423811_PM_at | - | - |
| Mouse Expression Set 430 | 1423811_at | - | - |
| Mouse Expression Set 430 | 1432488_a_at | - | - |
| Murine Genome U74v2 Set | 96029_at | - | - |
| Murine Genome U74v2 Set | 96029_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432488_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423811_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423811_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432488_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423811_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432488_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432488_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423811_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 460504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6660603 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6660603 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107000600 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613841 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232327 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613841 | - | - |
| mouse-6 v1.1 (Illumina) | scl24988.17.1_42-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660603 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107000600 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24988.17.1_42-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107000600 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2613841 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2613841 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2549077 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24988.17.1_42-S | - | - |

Sf3a3
Genomic Map