Assay Details
| Gene Symbol: Snx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029394.3 | NP_083670.1 | 2 - 3 | 273 | 87 | |
| GenBank mRNA | AK012711.1 | - | 2 - 3 | 165 | 87 | |
| AK017610.1 | - | 2 - 3 | 167 | 87 | ||
| BC051168.1 | - | 2 - 3 | 276 | 6306904 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx24
Genomic Map