| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1429669_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429669_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429669_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0800020819.mm.1 |
NR_026888.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020819.mm.1 |
NR_026888.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020819.mm.1 |
TC0800002720.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1429669_at |
- |
- |
| Mouse Expression Set 430 |
1429669_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1429669_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1429669_PM_at |
- |
- |