Assay Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029508.3 | NP_083784.1 | 6 - 7 | 731 | 100 | |
| GenBank mRNA | AK002387.1 | - | 6 - 7 | 754 | 100 | |
| AK017984.1 | - | 6 - 7 | 732 | 100 | ||
| AK146133.1 | - | 6 - 7 | 801 | 100 | ||
| BC139258.1 | - | 6 - 7 | 692 | 8862280 | 100 | |
| BC139259.1 | - | 6 - 7 | 692 | 8862281 | 100 | |
| BC145590.1 | - | 6 - 7 | 692 | 9054156 | 100 | |
| BC145591.1 | - | 6 - 7 | 689 | 9054157 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcgf5
Genomic Map