Assay Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029522.2 | NP_083798.2 | 1 - 2 | 264 | 77 | |
| GenBank mRNA | AK040996.1 | - | 1 - 2 | 153 | 77 | |
| AK143552.1 | - | 1 - 2 | 265 | 77 | ||
| AY081187.1 | - | 1 - 2 | 108 | 77 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK040996.1,AK143552.1,AY081187.1,NM_029522.2 transcripts.
Set Membership: |
|

Gpsm2
Genomic Map