Assay Details
| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029578.3 | NP_083854.3 | 6 - 7 | 591 | 67 | |
| GenBank mRNA | AK011439.1 | - | 5 - 6 | 479 | 67 | |
| AK011555.1 | - | 6 - 7 | 582 | 67 | ||
| AK154341.1 | - | 6 - 7 | 568 | 67 | ||
| AK165685.1 | - | 5 - 6 | 474 | 67 | ||
| AK165955.1 | - | 6 - 7 | 575 | 67 | ||
| BC021419.1 | - | 6 - 7 | 567 | 5012877 | 67 | |
| BY710811.1 | - | 6 - 7 | 579 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgds
Genomic Map