Assay Details
| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029578.3 | NP_083854.3 | 7 - 8 | 642 | 68 | |
| GenBank mRNA | AK011439.1 | - | 6 - 7 | 530 | 68 | |
| AK011555.1 | - | 7 - 8 | 633 | 68 | ||
| AK154341.1 | - | 7 - 8 | 619 | 68 | ||
| AK165685.1 | - | 6 - 7 | 525 | 68 | ||
| AK165955.1 | - | 7 - 8 | 626 | 68 | ||
| BY710811.1 | - | 7 - 8 | 630 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgds
Genomic Map