Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700003166.mm.1 |
NM_029624.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003166.mm.1 |
NM_029624.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003166.mm.1 |
NR_045520.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003166.mm.1 |
NR_045520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003166.mm.1 |
TC1700000380.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003167.mm.1 |
NM_029624.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003167.mm.1 |
NM_029624.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003167.mm.1 |
NR_045520.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003167.mm.1 |
NR_045520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003167.mm.1 |
TC1700000380.mm.1 |
MTA TC |