| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1429855_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429855_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429855_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
76705_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
76705_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0X00001632.mm.1 |
NR_033731.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001632.mm.1 |
NR_033731.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001632.mm.1 |
TC0X00000234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00001630.mm.1 |
NR_033731.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001630.mm.1 |
NR_033731.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001630.mm.1 |
TC0X00000234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00001631.mm.1 |
NR_033731.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001631.mm.1 |
NR_033731.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001631.mm.1 |
TC0X00000234.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1429855_at |
- |
- |
| Mouse Expression Set 430 |
1429855_PM_at |
- |
- |
| Murine Genome U74v2 Set |
166326_at |
- |
- |
| Murine Genome U74v2 Set |
166326_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1429855_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1429855_PM_at |
- |
- |