Assay Details
Gene Symbol: Rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122685.1 | NP_001116157.1 | 4 - 5 | 983 | 80 | |
NM_029777.3 | NP_084053.3 | 4 - 5 | 805 | 80 | ||
GenBank mRNA | AK015167.1 | - | 4 - 5 | 925 | 80 | |
AK029751.1 | - | 4 - 5 | 788 | 80 | ||
AK076751.1 | - | 4 - 5 | 984 | 80 | ||
AK079448.1 | - | 4 - 5 | 768 | 80 | ||
AK142277.1 | - | 4 - 5 | 794 | 80 | ||
AK161153.1 | - | 4 - 5 | 761 | 80 | ||
AY973322.1 | - | 3 - 4 | 656 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
