Assay Details
| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109913.1 | NP_001103383.1 | 8 - 9 | 736 | 88 | |
| NM_029804.3 | NP_084080.1 | 7 - 8 | 853 | 88 | ||
| GenBank mRNA | AK011521.1 | - | 7 - 8 | 799 | 88 | |
| AK133911.1 | - | 7 - 8 | 788 | 88 | ||
| AK145343.1 | - | 8 - 9 | 674 | 88 | ||
| AK159848.1 | - | 8 - 9 | 668 | 88 | ||
| AK166670.1 | - | 7 - 8 | 788 | 88 | ||
| AK168347.1 | - | 8 - 9 | 671 | 88 | ||
| AK169101.1 | - | 8 - 9 | 671 | 88 | ||
| AK171892.1 | - | 8 - 9 | 671 | 88 | ||
| AK220299.1 | - | 8 - 9 | 696 | 88 | ||
| BC017539.1 | - | 8 - 9 | 666 | 88 | ||
| BC035525.1 | - | 8 - 9 | 671 | 88 | ||
| BC054785.1 | - | 2255 | 88 | |||
| BC065172.1 | - | 8 - 9 | 666 | 6415170 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpm
Genomic Map