Assay Details
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029842.5 | NP_084118.1 | 6 - 7 | 1172 | 89 | |
GenBank mRNA | AK014006.1 | - | 6 - 7 | 1116 | 89 | |
BC024807.1 | - | 6 - 7 | 1100 | 5355270 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
