Assay Details
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029842.5 | NP_084118.1 | 7 - 8 | 1259 | 83 | |
GenBank mRNA | AK014006.1 | - | 7 - 8 | 1203 | 83 | |
BC024807.1 | - | 7 - 8 | 1187 | 5355270 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
chromatin modification
positive regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
chromatin binding
oxidoreductase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K36 specific)
G2/M transition of mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
chromatin modification
positive regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
chromatin binding
oxidoreductase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K36 specific)
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494602 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494600 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494603 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494598 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494601 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030060 | BC024807.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030060 | NM_029842.5 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030060 | AK014006.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66102 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856208 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130649 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33449 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8443 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184421 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751859 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99802 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972820 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913060 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453207 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872450 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64795 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115665 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160869 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102707 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030060 | BC024807.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030060 | NM_029842.5 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030060 | AK014006.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494596 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494587 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494574 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494591 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494579 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494588 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494572 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494611 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494573 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494576 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25494581 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102707 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160869 | - | - |
Whole Mouse Genome G4122A | A_51_P371408 | BC024807.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P371408 | NM_029842.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P371408 | AK014006.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113080 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136571 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102707 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160869 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06358082 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06358091 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06358073 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06358080 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494603 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494600 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494601 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494598 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25494602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25494656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06304190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06358092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06460851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06227577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06155303 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06304189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06569698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06396628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06358073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06358066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06109602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494574 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06549475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06358071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06109604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06423848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06358091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06549471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06463531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06050834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06423849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05973771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25494573 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10557318 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557321 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557320 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557324 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557325 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557319 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557323 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10557322 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451384_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10557317 | - | - |
Mouse Genome 430 2.0 Array | 1451384_at | - | - |
Mouse Genome 430 2.0 Array | 1451384_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77035_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77035_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700015230.mm.1 | TC0700001791.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700015230.mm.1 | NM_029842.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700015230.mm.1 | NM_029842.5 | Refseq |
Mouse Expression Set 430 | 1451384_PM_at | - | - |
Mouse Expression Set 430 | 1451384_at | - | - |
Murine Genome U74v2 Set | 165675_at | - | - |
Murine Genome U74v2 Set | 165675_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451384_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451384_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451384_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451384_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 702416 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6660575 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6660575 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006660575 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244329 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244329 | - | - |
mouse-6 v1.1 (Illumina) | scl32071.8.1_31-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6660575 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32071.8.1_31-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660575 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244329 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2821736 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2821736 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244329 | - | - |
