Assay Details
Gene Symbol: Cep83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029852.2 | NP_084128.2 | 6 - 7 | 1150 | 111 | |
GenBank mRNA | AK015007.1 | - | 6 - 7 | 1287 | 111 | |
AK141627.1 | - | 6 - 7 | 1271 | 111 | ||
BC028960.1 | - | 5 - 6 | 851 | 111 | ||
BC043468.1 | - | 5 - 6 | 764 | 111 | ||
BC070464.1 | - | 6 - 7 | 1150 | 30608497 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
