| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00014185.mm.1 |
NM_001166365.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00014185.mm.1 |
NM_001166365.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00014185.mm.1 |
NM_001166583.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00014185.mm.1 |
NM_001166583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00014185.mm.1 |
NM_030167.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00014185.mm.1 |
NM_030167.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00014185.mm.1 |
TC0X00002236.mm.1 |
MTA TC |