Assay Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030225.4 | NP_084501.1 | 3 - 4 | 212 | 115 | |
| GenBank mRNA | AK019713.2 | - | 209 | 115 | ||
| AK054053.1 | - | 3 - 4 | 166 | 115 | ||
| AK149664.1 | - | 3 - 4 | 188 | 115 | ||
| AK158877.1 | - | 3 - 4 | 213 | 115 | ||
| AK168570.1 | - | 3 - 4 | 188 | 115 | ||
| AK169943.1 | - | 3 - 4 | 166 | 115 | ||
| BC006702.1 | - | 3 - 4 | 149 | 3583936 | 115 | |
| CT010197.1 | - | 3 - 4 | 148 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlst
Genomic Map