Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000000126.mm.1 |
NM_031185.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000126.mm.1 |
NM_031185.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000126.mm.1 |
TC1000000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000123.mm.1 |
NM_031185.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000123.mm.1 |
NM_031185.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000123.mm.1 |
TC1000000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000125.mm.1 |
NM_031185.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000125.mm.1 |
NM_031185.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000125.mm.1 |
TC1000000021.mm.1 |
MTA TC |