Assay Details
| Gene Symbol: Rab15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134050.4 | NP_598811.3 | 4 - 5 | 531 | 89 | |
| GenBank mRNA | AB232610.1 | - | 4 - 5 | 324 | 89 | |
| AK158637.1 | - | 4 - 5 | 453 | 89 | ||
| AK158768.1 | - | 4 - 5 | 485 | 89 | ||
| BC013790.1 | - | 4 - 5 | 477 | 2655151 | 89 | |
| BC027769.1 | - | 4 - 5 | 531 | 5345297 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab15
Genomic Map