Assay Details
| Gene Symbol: Ddx41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134059.2 | NP_598820.2 | 7 - 8 | 790 | 70 | |
| GenBank mRNA | AK151427.1 | - | 7 - 8 | 664 | 70 | |
| AK156957.1 | - | 7 - 8 | 658 | 70 | ||
| AK168043.1 | - | 7 - 8 | 663 | 70 | ||
| BC011308.1 | - | 7 - 8 | 644 | 3256762 | 70 | |
| BC023711.1 | - | 7 - 8 | 665 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx41
Genomic Map