Assay Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134078.4 | NP_598839.2 | 6 - 7 | 1034 | 75 | |
| GenBank mRNA | BB621791.1 | - | 2 - 3 | 167 | 75 | |
| BC024115.1 | - | 6 - 7 | 1022 | 5100379 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chmp7
Genomic Map